Order Entry
Austria
ContactUsLinkComponent
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide
Health
Irritant
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide
Catalog # MOLE14706101-5G
Supplier:  Molekula
CAS Number:  
Health
Irritant
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide
Catalog # MOLE14706101-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :414.33 g/mol
EINECS:206-069-5
 :GHS08
 :GHS07
Storage Temperature:Refrigerator
CAS Number:298-93-1
Formula:C₁₈H₁₆N₅BrS
MDL Number:MFCD00011964