

3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:298-93-1
:
Formula:C₁₈H₁₆N₅BrS
MDL Number:MFCD00011964
:414.33 g/mol
EINECS:206-069-5
:GHS08
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together