Order Entry
Austria
ContactUsLinkComponent
 
DL-α-Tocopherol acetate
Catalog # ROTH3805.1
Supplier:  Roth Carl
CAS Number:  
DL-α-Tocopherol acetate
Catalog # ROTH3805.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :472.75 g/mol
EINECS:231-710-0
CAS Number:7695-91-2
 :52225-20-4
Formula:C₃₁H₅₂O₃
MDL Number:MFCD00072042