Order Entry
Austria
ContactUsLinkComponent
all-rac-α-Tocopheryl acetate (Vitamin E acetate), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
all-rac-α-Tocopheryl acetate (Vitamin E acetate), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Certificates
all-rac-α-Tocopheryl acetate (Vitamin E acetate), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Certificates

Specification Test Results

  • Assay (GC, Ph. Eur.):
    96.5 - 102.0 %
  • Assay (GC, USP):
    96.0 - 102.0 %
  • Assay (GC, FCC):
    960 - 1020 I.U./g
  • Identity (IR):
    conforms
  • Optical rotation α 20/D (100 g/l; anhydrous ethanol):
    -0.01 - 0.01 °
  • As (Arsenic):
    ≤ 0.0003 %
  • Cu (Copper):
    ≤ 0.0025 %
  • Pb (Lead):
    ≤ 0.0002 %
  • Zn (Zinc):
    ≤ 0.0010 %
  • Acetic acid (HS-GC):
    ≤ 5000 ppm
  • Heptane (HS-GC):
    ≤ 5000 ppm
  • Methanol (HS-GC):
    ≤ 3000 ppm
  • Pyridine (HS-GC):
    ≤ 200 ppm
  • Toluene (HS-GC):
    ≤ 890 ppm
  • Other residual solvents (ICH Q3C):
    excluded by manucfacturing process
  • Related substances (Ph. Eur., impurity A):
    ≤ 0.5 %
  • Related substances (Ph. Eur., impurity B):
    ≤ 1.5 %
  • Related substances (Ph. Eur., impurity C):
    ≤ 0.5 %
  • Related substances (Ph. Eur., sum of impurities D and E):
    ≤ 1.0 %
  • Related substances (Ph. Eur., any other impurity):
    ≤ 0.25 %
  • Related substances (Ph. Eur., total impurities):
    ≤ 2.5 %
  • Acidity:
    conforms

Specification Test Results

Related Information

Frequently Bought Together