
Diethyl dibromomalonate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:631-22-1
:
Formula:C₇H₁₀Br₂O₄
MDL Number:MFCD00015154
:317.96 g/mol
Boiling Pt:108…110 °C (4 mmHg)
Density:1.680
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:211-151-9
UN:1760
ADR:8,III
:GHS05
Related Information
Frequently Bought Together