Order Entry
Austria
ContactUsLinkComponent
Ammonium acetate 5.0 M in aqueous solution
Ammonium acetate 5.0 M in aqueous solution
Catalog # ROCKMB-041
CAS Number:  
undefined
Ammonium acetate 5.0 M in aqueous solution
Catalog # ROCKMB-041
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :77.08 g/mol
Storage Temperature:Ambient
EINECS:211-162-9
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066