

Erythrosin B disodium salt ≥80%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:16423-68-0
:568-63-8
Formula:C₂₀H₆I₄Na₂O₅
MDL Number:MFCD00144257
:879.86 g/mol
:M-Clarity™ quality level = MQ100
Flash Pt:182 °C (closed cup)
EINECS:240-474-8
:GHS07
:GHS09
Specification Test Results
- Appearance (colour):Red to very dark red and red-brown and very dark red-brown
- Appearance (form):Powder or crystals
- Assay (gravimetr.):≥80%
- Thin layer chromatogr.:Corresponds to standard
- Solubility (colour):Red to very dark red
- Solubility (turbidity):Report result
- Solubility (method):0.1 g in 10 ml water
- Loss on drying:≤10%
- Wavelength (1) (UV):520 - 530 nm
- Extinction (1%/1 cm) (1) (UV):≥800
- Solvent (UV):H₂O+1 ml 1% sodium carbonate
Specification Test Results
Frequently Bought Together