DL-Threonine ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:80-68-2
:
Formula:C₄H₉NO₃
MDL Number:MFCD00063722
Melting Pt:238…248 °C
:119.12 g/mol
Storage Temperature:Ambient
EINECS:201-300-6
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- Loss on Drying:≤0.2%
- Optical Rotation:-1° to +1° (c=3, water)
Specification Test Results
Frequently Bought Together