Organic reference standard, Acenaphthylene 5000 µg/ml in methanol
Catalog # ACCUASE0075
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:208-96-8
- Description:Acenaphthylene 5,000 µg/ml in methanol
- Formula:C₁₂H₈
- Description:Acenaphthylene