3-Methoxyphenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:150-19-6
:
Formula:C₇H₈O₂
MDL Number:MFCD00002267
Boiling Pt:244 °C (760 mmHg)
Melting Pt:–17 °C
Density:1.145 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:205-754-6
ADR:6.1,III
:124.14 g/mol
UN:2810
:GHS05
:GHS06
Specification Test Results
- Appearance:Clear pink-red to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5510 to 1.5530 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together