ContactUsLinkComponent
Measuring beakers, conical, graduated
Measuring beakers, conical, graduated
Catalog # BOCH8650
Supplier:  BOCHEM
Supplier #: 8650
undefined
Measuring beakers, conical, graduated
Catalog # BOCH8650
Supplier:  BOCHEM
Supplier #: 8650
Log in for availability
Select an Option
125 mm
Select an Option
500 ml
Select an Option
95 mm
Select an Option
1
About this item
Stainless steel, 18/10, with handle and spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Frequently Bought Together