Order Entry
Austria
Orders LinkContactUsLinkComponent
Digital burettes, Technotrate

Digital burettes, Technotrate

0 - 50 ml, 0,01 ml, ≤0,2 %, ≤0,1 %
Catalog # KART83250
Supplier:   KARTELL
Digital burettes, Technotrate
Catalog # KART83250
Supplier:  KARTELL
Digital burettes, Technotrate
Digital burettes, Technotrate

0 - 50 ml, 0,01 ml, ≤0,2 %, ≤0,1 %

Price
2 340,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Digital burette to cover the volume range of 0 to 50 ml. By pushing the key 'DOS' the user can preselect the burette volume for titrations. By pressing the buttons 'ZERO' and 'TITR' the titration can be started immediately. For liquids of different viscosities/densities the TECHNOTRATE can be easily recalibrated by pushing the button 'O/CAL'.

  • Possible to print the daily/weekly values in connection to a computer and printer by using the interface 'Witolink'
  • 'Air purging' without problems or loss of reagent/titration medias
  • No more spills, safe and secure restposition
  • Automatic power-cut off after three minutes
  • Rechargeable battery

Documents

Specifications
  • Capacity:
    0 - 50 ml
  • Accuracy:
    ≤0,2 %
  • Division:
    0,01 ml
  • Imprecision:
    ≤0,1 %
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Bottle, amber, GL 45, 1000 ml
    Bottle, amber, GL 45, 1000 ml
    KART83011
    Log in for availability
    1 * 1 ST - 48,30 €

Product Information

  • Catalog #
    Availability
    Price
  • Bottle, amber, GL 45, 2500 ml
    Bottle, amber, GL 45, 2500 ml
    KART83012
    Log in for availability
    1 * 1 ST - 95,60 €

Product Information

  • Catalog #
    Availability
    Price
  • Suction tube, Dosipet, 2,5 - 10 ml
    Suction tube, Dosipet, 2,5 - 10 ml
    KART83016
    Log in for availability
    1 * 1 ST - 41,60 €

Product Information

  • Catalog #
    Availability
    Price
  • Discharge tube, 220 V-50Hz
    Discharge tube, 220 V-50Hz
    KART83019
    Log in for availability
    1 * 1 ST - 35,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Discharge tube, (FEP), DOSIPET-Technotrate, 2,5 - 100 ml
    Discharge tube, (FEP), DOSIPET-Technotrate, 2,5 - 100 ml
    KART83020
    Log in for availability
    1 * 1 ST - 41,70 €

Product Information

  • Catalog #
    Availability
    Price
  • Set of 5 adapters: A25, A28, A38, A40, A45
    Set of 5 adapters: A25, A28, A38, A40, A45
    KART83023
    Log in for availability
    1 * 1 ST - 63,80 €

Product Information

  • Catalog #
    Availability
    Price
  • Glass non return valve for checking, DOSIPET-Technotrate
    Glass non return valve for checking, DOSIPET-Technotrate
    KART83026
    Log in for availability
    1 * 1 ST - 141,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Calcium chloride tube
    Calcium chloride tube
    KART8302400
    Log in for availability
    1 * 1 ST - 74,20 €

Product Information

  • Catalog #
    Availability
    Price
  • Bottle, amber, GL 32, 250 ml
    Bottle, amber, GL 32, 250 ml
    KART8302700
    Log in for availability
    1 * 1 ST - 44,90 €

Product Information

  • Catalog #
    Availability
    Price
  • Bottle, amber, GL 32, 500 ml
    Bottle, amber, GL 32, 500 ml
    KART8302800
    Log in for availability
    1 * 1 ST - 46,80 €