Orders LinkContactUsLinkComponent
Test tube racks, rectangular
Test tube racks, rectangular
Supplier:   BOCHEM
Test tube racks, rectangular
Supplier:  BOCHEM
Certificates
Test tube racks, rectangular
Test tube racks, rectangular
5 Options Available Below
Product Details & Documents
Rack, made of stainless steel 18/10.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Product Family Options
Product Information
  • No. of Holes
    For tube Ø
    W×D×H
    Availability
    Price
  • 12 (2×6)
    17 mm
    145×52×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,90 €
    33,90 €
    1 * 1 ST
       33,90 €
  • 24 (2×12)
    17 mm
    278×52×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,90 €
    36,90 €
    1 * 1 ST
       36,90 €
  • 48 (4×12)
    17 mm
    287×97×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,40 €
    64,40 €
    1 * 1 ST
       64,40 €
  • 12 (2×6)
    20 mm
    164×60×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,40 €
    43,40 €
    1 * 1 ST
       43,40 €
  • 24 (2×12)
    20 mm
    310×60×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,40 €
    56,40 €
    1 * 1 ST
       56,40 €