Orders LinkContactUsLinkComponent
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
Supplier:   THOMSON INSTRUMENT
Filter vials, SINGLE StEP™
Supplier:  THOMSON INSTRUMENT
Certificates
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
14 Options Available Below
Product Details & Documents
Simple replacements for HPLC vials, HPLC caps, syringes, needles, and syringe filters. Filter vials minimise steps and loss of product. They protect HPLC, GC, GC-MS and LC-MS systems against contaminants and particulates.
  • Compatible with both UHPLC and standard HPLC autosamplers
  • Colour coded pre-slit caps for easy identification of membrane and porosity
  • Low dead volume unit contains rugged PP insert with 450 µl loading capacity
Product Family Options
Product Information
  • Membrane
    Pore size
    Colour
    Availability
    Price
  • Nylon
    0,2 µm
    Black
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 742,00 €
    742,00 €
    1 * 200 ST
       742,00 €
  • Nylon
    0,2 µm
    Black
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 860,00 €
    1 860,00 €
    1 * 500 ST
       1 860,00 €
  • PES
    0,2 µm
    Grey
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 742,00 €
    742,00 €
    1 * 200 ST
       742,00 €
  • PES
    0,2 µm
    Grey
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 860,00 €
    1 860,00 €
    1 * 500 ST
       1 860,00 €
  • PTFE
    0,2 µm
    Green
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 742,00 €
    742,00 €
    1 * 200 ST
       742,00 €
  • PTFE
    0,2 µm
    Green
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 860,00 €
    1 860,00 €
    1 * 500 ST
       1 860,00 €
  • PVDF
    0,2 µm
    Red
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 742,00 €
    742,00 €
    1 * 200 ST
       742,00 €
  • PVDF
    0,2 µm
    Red
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 1 860,00 €
    1 860,00 €
    1 * 100 ST
       1 860,00 €
  • Nylon
    0,45 µm
    Pink
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 742,00 €
    742,00 €
    1 * 200 ST
       742,00 €
  • Nylon
    0,45 µm
    Pink
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 860,00 €
    1 860,00 €
    1 * 500 ST
       1 860,00 €
  • PTFE
    0,45 µm
    Blue
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 742,00 €
    742,00 €
    1 * 200 ST
       742,00 €
  • PTFE
    0,45 µm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 860,00 €
    1 860,00 €
    1 * 500 ST
       1 860,00 €
  • PVDF
    0,45 µm
    Yellow
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 742,00 €
    742,00 €
    1 * 200 ST
       742,00 €
  • PVDF
    0,45 µm
    Yellow
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 860,00 €
    1 860,00 €
    1 * 500 ST
       1 860,00 €