Laboratory tables, with ground plate
Supplier: BOCHEM
Certificates
About this item
These laboratory tables are made up of 18/10 stainless steel and height adjustable.
- Drawer face with user friendly handle bar
- Noise protection
Working plate with matt finishing and corners are continuously welded and polished.
Drawer with guard rails made of stainless steel to mount GN-drawer. Drawers can be taken off for cleaning, but are secured against drawing out.
Frequently Bought Together