Order Entry
Austria
Orders LinkContactUsLinkComponent
Spoon spatulas, analyse type

Spoon spatulas, analyse type

235 mm, 25×18 mm
Catalog # BOCH3216
Supplier:   BOCHEM
Spoon spatulas, analyse type
Catalog # BOCH3216
Supplier:  BOCHEM
Spoon spatulas, analyse type
Spoon spatulas, analyse type

235 mm, 25×18 mm

Price
4,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Spoon spatula with one spoon end and one flat end. Spoon size: 25×18 mm

Specifications
  • Length:
    235 mm
  • Spoon size:
    25×18 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.