Orders LinkContactUsLinkComponent
Separating funnels, conical, PMP
Separating funnels, conical, PMP
500 ml, 29/32 NS
Catalog # KART978
Supplier:   KARTELL
Separating funnels, conical, PMP
Catalog # KART978
Supplier:  KARTELL
Separating funnels, conical, PMP
Separating funnels, conical, PMP
500 ml, 29/32 NS
Price
115,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Separating funnels, made of PMP, crystal clear, with permanent graduations.
  • Needle control for precise control of flow
  • Top unscrews and valve can be detached for thorough cleaning
  • Autoclavable and chemical resistant
  • Complete with stopper
Documents
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    500 ml