Orders LinkContactUsLinkComponent
Scintillation vials
Scintillation vials
Supplier:   Simport Scientific
Scintillation vials
Supplier:  Simport Scientific
Certificates
Scintillation vials
Scintillation vials
2 Options Available Below
Product Details & Documents
These scintillation vials are made of PP. Caps are made of HDPE.
  • Easy removal of caps (¼ turn-twist), leakproof
  • High degree of resistance to organic solvents
  • Shoulderless vial features a full width opening of 23 mm for ease of access
  • Sufficient translucence so that reagent levels or the presence of filter paper can be easily detected
The quality of the sealing system is such that it is not necessary to tighten the closure with a lot of pressure to achieve a leakproof seal. The HDPE closure may not be autoclaved.
Product Family Options
Product Information
  • Capacity
    Ø×H
    Cap included
    Packed
    Availability
    Price
  • 20,00 ml
    26×61 mm
    Yes
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 190,00 €
    190,00 €
    1 * 500 ST
       190,00 €
  • 20,00 ml
    26×61 mm
    Yes
    Tray/100
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 398,00 €
    398,00 €
    1 * 500 ST
       398,00 €