Order Entry
Austria
Orders LinkContactUsLinkComponent
3,5-Dinitroaniline

3,5-Dinitroaniline

Glass bottle for solids, 5 g
Catalog # MOLE34717589-5G
Marketsource
Supplier:   Molekula
3,5-Dinitroaniline
Catalog # MOLE34717589-5G
Supplier:  Molekula
Marketsource
CAS Number:  618-87-1
3,5-Dinitroaniline

Glass bottle for solids, 5 g

CAS Number:  618-87-1
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:618-87-1
  • Formula:C₆H₅N₃O₄
  • MDL Number:MFCD00007263
  • Molecular Weight:183.12 g/mol
  • Melting Pt:163 °C
  • Density:1.601 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:210-567-8
  • UN:1596
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications