Orders LinkContactUsLinkComponent
Laboratory jacks
 :   BOCHEM
Laboratory jacks
 :  BOCHEM
Laboratory jacks
Laboratory jacks
Aluminium or anodised aluminium. These lab jacks feature an improved scissor technology with reduced tolerances. The scissor construction is made of 18/10 stainless steel.
  • Ergonomic knob, rounded and user friendly
  • Smaller models up to 240×240 mm have a simple adjusting wheel, which can be operated by hand
  • Bigger models from 300×300 mm are recommended to be used in combination with a ratchet, which connects with a special adjusting wheel (ratchet included)
 : DIN 12897
Product Family Options
Aluminium
  • Plate size
    Max. static load capacity
    Max. dynamic load capacity
    Working height
    Colour
    With
  • 100×100 mm
    10 kg
    5 kg
    55 - 120 mm
    Green
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 179,00 €
    179,00 €
    1 * 1 ST
       179,00 €
  • 160×130 mm
    30 kg
    7 kg
    60 - 275 mm
    Green
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 146,00 €
    146,00 €
    1 * 1 ST
       146,00 €
  • 200×200 mm
    30 kg
    7 kg
    60 - 275 mm
    Green
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
    1 * 1 ST
       169,00 €
  • 240×240 mm
    30 kg
    7 kg
    60 - 275 mm
    Green
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €
  • 300×300 mm
    60 kg
    12 kg
    90 - 470 mm
    Green
    Adjusting wheel for ratchet
    1 * 1 ST
    1 * 1 ST - 602,00 €
    602,00 €
    1 * 1 ST
       602,00 €
  • 400×400 mm
    60 kg
    15 kg
    90 - 470 mm
    Green
    Adjusting wheel for ratchet
    1 * 1 ST
    1 * 1 ST - 850,00 €
    850,00 €
    1 * 1 ST
       850,00 €
  • 160×130 mm
    30 kg
    7 kg
    75 - 400 mm
    Green
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 193,00 €
    193,00 €
    1 * 1 ST
       193,00 €
  • 200×200 mm
    30 kg
    7 kg
    75 - 400 mm
    Green
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 216,00 €
    216,00 €
    1 * 1 ST
       216,00 €
Anodised aluminium
  • Plate size
    Max. static load capacity
    Max. dynamic load capacity
    Working height
    Colour
    With
  • 100×100 mm
    10 kg
    5 kg
    55 - 120 mm
    Blue
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 192,00 €
    192,00 €
    1 * 1 ST
       192,00 €
  • 240×240 mm
    30 kg
    7 kg
    60 - 275 mm
    Blue
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 211,00 €
    211,00 €
    1 * 1 ST
       211,00 €
  • 300×300 mm
    60 kg
    12 kg
    90 - 470 mm
    Blue
    Adjusting wheel for ratchet
    1 * 1 ST
    1 * 1 ST - 674,00 €
    674,00 €
    1 * 1 ST
       674,00 €
  • 400×400 mm
    60 kg
    15 kg
    90 - 470 mm
    Blue
    Adjusting wheel for ratchet
    1 * 1 ST
    1 * 1 ST - 950,00 €
    950,00 €
    1 * 1 ST
       950,00 €
  • 200×200 mm
    30 kg
    7 kg
    60 - 275 mm
    Blue
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €
  • 160×130 mm
    30 kg
    7 kg
    60 - 275 mm
    Blue
    Adjusting wheel
    1 * 1 ST
    1 * 1 ST - 160,00 €
    160,00 €
    1 * 1 ST
       160,00 €
Accessories
  •  
    Description
  • 245-0026
    Ratchet for laboratory jacks, length 190 mm
    1 * 1 ST - 47,60 €