4-Nitroaniline 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-01-6
:
Formula:C₆H₆N₂O₂
MDL Number:MFCD00007858
:138.13 g/mol
Merck Index:12,06681
Boiling Pt:332 °C (760 mmHg)
Melting Pt:146…151 °C
Density:1,24 g/cm³ (20 °C)
Flash Pt:199 °C
Storage Temperature:Ambient
EINECS:202-810-1
UN:1661
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:Yellow to brown Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:147 to 151 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together