Orders LinkContactUsLinkComponent
Sample containers with screw cap
Sample containers with screw cap
Supplier:   KARTELL
Sample containers with screw cap
Supplier:  KARTELL
Certificates
Sample containers with screw cap
Sample containers with screw cap
4 Options Available Below
Product Details & Documents
Made of PS, with PE screw cap and spoon.
  • Disposable
  • Available sterile or non sterile
  • Good chemical resistance
  • Lightweight
Product Family Options
Product Information
  • Capacity
    Type
    Cap colour
    Ext.Ø
    Height
    Packed
    Availability
    Price
  • 30 ml
    Sterile, with white spoon and label
    White
    25 mm
    90 mm
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 317,00 €
    317,00 €
    1 * 400 ST
       317,00 €
  • 30 ml
    Non sterile, with white spoon
    White
    25 mm
    90 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 239,00 €
    239,00 €
    1 * 400 ST
       239,00 €
  • 60 ml
    Non sterile, with white spoon
    Red
    38 mm
    65 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 299,00 €
    299,00 €
    1 * 500 ST
       299,00 €
  • 60 ml
    Sterile, with white spoon
    Red
    38 mm
    65 mm
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 286,00 €
    286,00 €
    1 * 400 ST
       286,00 €