Order Entry
Austria
Orders LinkContactUsLinkComponent
Spoons, PharmaSpoon

Spoons, PharmaSpoon

650 mm, 500 ml
Catalog # 300-0075
Supplier:   BURKLE
Spoons, PharmaSpoon
Catalog # 300-0075
Supplier:  BURKLE
Spoons, PharmaSpoon
Spoons, PharmaSpoon

650 mm, 500 ml

Price
603,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel V4A (1.4404), high polish finish. For taking samples of powdery substances, intermediate products and material in the pharmaceutical industry.

  • Possible residues are even visible in the trace sector (µg sector)
  • Also suitable for sterile sampling
  • Made of materials with excellent surface qualities to prevent cross-contamination
Specifications
  • Capacity:
    500 ml
  • Length:
    650 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.