Orders LinkContactUsLinkComponent
Process pH combination electrodes
Process pH combination electrodes
Process pH combination electrodes
Process pH combination electrodes
Product Family Options
  • Description
    Type
    pH
    Temp.
    Material
    Electrolyte
    Diaphr.
    Ø×L
    Connection
  • pH combination electrode, with sphere membrane
    L 1181 HD
    0 - 12
    −30...+80 °C
    Glass
    -
    -
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 277,00 €
    277,00 €
    1 * 1 ST
       277,00 €
  • pH combination electrode, with sphere membrane
    L 9080
    0 - 12
    −30...+80 °C
    Glass
    L 200
    3×Ceramic
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 441,00 €
    441,00 €
    1 * 1 ST
       441,00 €
  • pH combination electrode, with cylinder membrane
    H 7381 HD
    2 - 13
    −5...+100 °C
    Glass
    Gel
    Ceramic
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 306,00 €
    306,00 €
    1 * 1 ST
       306,00 €
  • pH combination electrode, with sphere membrane
    H 6281
    1 - 14
    0...+135 °C
    Glass
    KCl 3 mol/l
    Ceramic
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 326,00 €
    326,00 €
    1 * 1 ST
       326,00 €
  • pH combination electrode, with sphere membrane
    H 9080
    1 - 14
    0...+135 °C
    Glass
    KCl 3 mol/l
    Ceramic
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 404,00 €
    404,00 €
    1 * 1 ST
       404,00 €
  • pH combination electrode, with sphere membrane
    S 9080
    2 - 14
    +10...+135 °C
    Glass
    KCl 3 mol/l
    Ceramic
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 465,00 €
    465,00 €
    1 * 1 ST
       465,00 €
  • pH combination electrode, with cylinder membrane and PT1000
    H 7381-T1000 VP
    2 - 13
    −5...+80 °C
    Glass
    Gel
    Ceramic
    12×120 mm
    VP plug head
    1 * 1 ST
    1 * 1 ST - 387,00 €
    387,00 €
    1 * 1 ST
       387,00 €
  • pH combination electrode, with sphere membrane
    H 6181 HD
    0 - 14
    0...+135 °C
    Glass
    KCl 3 mol/l
    Ceramic
    12×170 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 331,00 €
    331,00 €
    1 * 1 ST
       331,00 €
  • pH combination electrode, with sphere membrane
    H 6381
    1 - 14
    0...+135 °C
    Glass
    KCl 3 mol/l
    Ceramic
    12×320 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 359,00 €
    359,00 €
    1 * 1 ST
       359,00 €
  • pH combination electrode, with sphere membrane
    H 9180
    1 - 14
    0...+135 °C
    Glass
    KCl 3 mol/l
    Ground joint
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 462,00 €
    462,00 €
    1 * 1 ST
       462,00 €
  • pH combination electrode, with sphere membrane
    S 9180
    2 - 14
    +10...+135 °C
    Glass
    KCl 3 mol/l
    Ground joint
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 529,00 €
    529,00 €
    1 * 1 ST
       529,00 €
  • pH combination electrode, with sphere membrane
    L 9180
    0 - 12
    −30...+80 °C
    Glass
    L 200
    Ground joint
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 491,00 €
    491,00 €
    1 * 1 ST
       491,00 €
  • pH-combination electrodes with sphere membrane
    H 64
    0 - 14
    0...+100 °C
    Glass
    KCl 3 mol/l
    Ground joint
    12×170 mm
    Plug head
    1 * 1 ST
    1 * 1 ST - 386,00 €
    386,00 €
    1 * 1 ST
       386,00 €
  • pH combination electrode, with dome membrane
    H 8281 HD
    2 - 13
    0...+100 °C
    Glass
    Referid®
    KPG annular gap
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 311,00 €
    311,00 €
    1 * 1 ST
       311,00 €
  • pH combination electrode, with dome membrane
    H 8181 HD
    2 - 13
    0...+100 °C
    Glass
    Referid®
    KPG annular gap
    12×170 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • pH combination electrode, with dome membrane
    H 8481 HD
    1 - 14
    0...+100 °C
    Glass
    Referid®
    KPG annular gap
    12×225 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 367,00 €
    367,00 €
    1 * 1 ST
       367,00 €
  • pH combination electrode, with dome membrane
    H 8381 HD
    2 - 13
    0...+100 °C
    Glass
    Referid®
    KPG annular gap
    12×320 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 408,00 €
    408,00 €
    1 * 1 ST
       408,00 €
  • pH combination electrode, with sphere membrane
    H 6291
    1 - 14
    0...+135 °C
    Glass
    KCl 3 mol/l
    Platinum
    12×120 mm
    Screw plug head
    1 * 1 ST
    1 * 1 ST - 334,00 €
    334,00 €
    1 * 1 ST
       334,00 €