Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stoppers
PE/LDPE/HDPE Stoppers
Stoppers for test tubes
Stoppers for test tubes
Stoppers for test tubes
Catalog # 212-0222
Supplier:
KARTELL
Expand image
Stoppers for test tubes
Catalog # 212-0222
Supplier:
KARTELL
Specifications
Ø bottom:
11 mm
Colour:
Natural
Pk:
1.000
Specifications
About this item
Product Details
Stoppers for disposible tubes, PE.
Product Details