Powder samplers, MicroSampler
About this item
Stainless steel AISI 316 L (1,4404). The MicroSampler has been specially designed for the sampling of small quantities of powder.
- Point sampling
- Various lengths and volumes are available
- Tip can be removed for easy cleaning
- Additional tips are available in different sizes
- Sterilisable
Volumes of 0,2 ml, 0,5 ml, 1,0 ml, 2,0 ml, 5,0 ml, and 10 ml can be sampled easily, quickly, and cleanly from different sampling depths from sacks or containers.
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.
Ordering information: Please order lance and head separately.
13 Options Available Below
Product Details & Documents
Stainless steel AISI 316 L (1,4404). The MicroSampler has been specially designed for the sampling of small quantities of powder.
- Point sampling
- Various lengths and volumes are available
- Tip can be removed for easy cleaning
- Additional tips are available in different sizes
- Sterilisable
Volumes of 0,2 ml, 0,5 ml, 1,0 ml, 2,0 ml, 5,0 ml, and 10 ml can be sampled easily, quickly, and cleanly from different sampling depths from sacks or containers.
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.
Ordering information: Please order lance and head separately.
Product Family Options
MicroSampler heads
Product Information
MicroSampler (without head)
Product Information
Accessories
Product Information
- Catalog #MaterialVolumeAvailabilityPrice
- Head for Ø 12 mm tube
Product Information
- Catalog #MaterialVolumeAvailabilityPrice
- Head for Ø 25 mm tubeHead for Ø 25 mm tube331-0025Stainless steel 316L2,0 mlLog in for availability1 * 1 ST - 112,00 €
Product Information
- Catalog #MaterialVolumeAvailabilityPrice
- Head for Ø 12 mm tube
Product Information
- Catalog #MaterialVolumeAvailabilityPrice
- Head for Ø 25 mm tubeHead for Ø 25 mm tube331-2192Stainless steel 316L5,0 mlLog in for availability1 * 1 ST - 112,00 €
Product Information
- Catalog #MaterialVolumeAvailabilityPrice
- Head for Ø 12 mm tube
Product Information
- Catalog #MaterialVolumeAvailabilityPrice
- Head for Ø 25 mm tube