Order Entry
Austria
ContactUsLinkComponent
 
3-Iodobenzonitrile 97%
 
undefined
3-Iodobenzonitrile 97%

 

  • Appearance:
    White to brown Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 44 °C
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together