1,3,5-Trifluorobenzene
Catalog # MOLEM36685457
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:132.09 g/mol
Boiling Pt:75 °C (1013 hPa)
Density:1.277 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-751-2
UN:2387
ADR:3,II
CAS Number:372-38-3
Formula:C₆H₃F₃
MDL Number:MFCD00000333