Orders LinkContactUsLinkComponent
Samplers, PowderProof®
Samplers, PowderProof®
Supplier:   BURKLE
Samplers, PowderProof®
Supplier:  BURKLE
Certificates
Samplers, PowderProof®
16 Options Available Below
Product Details & Documents
Quality assurance starts with the inspection of incoming goods. An all-layer sample of bulk goods is no longer a problem with PowderProof®, the sampler for powders and granules.
  • Many uses
  • Stainless steel AISI 316L (1.4404)/AISI 301 (1.4310)
  • Includes motor, two rechargeable batteries, fast charger, lance, spiral screw, bags, bag clip, and cleaning brush
  • Additional spiral screws available as accessory
PowderProof® is the practically proven professional solution for mobile sampling of powders and granules. Different lances and augers can be attached to the robust drive unit and swapped without any problem. This modular design makes PowderProof® a versatile instrument for sampling powders and granules from drums, barrels, bags, or sacks.
Safe and reliable sampling is made even easier with useful accessories such as basket-enclosed bottles, sample bags, and close-it control seals.
PowderProof® with lance for horizontal use, such as sampling from sacks, with lance for vertical use, for sampling from open containers such as barrels, drums, or bags.
Product Family Options
PowderProof®
Product Information
  • Length
    Ø
    Availability
    Price
  • 300-0193
    Sampler, stainless steel V4A, with horizontal lance, for powders up to 2 mm Ø
    Sampler, stainless steel V4A, with horizontal lance, for powders up to 2 mm Ø
    300 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 180,00 €
    2 180,00 €
    1 * 1 ST
       2 180,00 €
  • 300-0195
    Sampler, stainless steel V4A, with vertical lance, for powders up to 2 mm Ø
    Sampler, stainless steel V4A, with vertical lance, for powders up to 2 mm Ø
    600 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 340,00 €
    2 340,00 €
    1 * 1 ST
       2 340,00 €
  • 300-0017
    Sampler, stainless steel V4A, with vertical lance, for powders up to 2 mm Ø
    Sampler, stainless steel V4A, with vertical lance, for powders up to 2 mm Ø
    900 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 450,00 €
    2 450,00 €
    1 * 1 ST
       2 450,00 €
  • 300-0198
    Sampler, stainless steel V4A, with vertical lance, for powders up to 2 mm Ø
    Sampler, stainless steel V4A, with vertical lance, for powders up to 2 mm Ø
    1200 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 720,00 €
    2 720,00 €
    1 * 1 ST
       2 720,00 €
  • 300-0192
    Sampler, stainless steel V4A, with horizontal lance, for granules from 2 up to 4 mm Ø
    Sampler, stainless steel V4A, with horizontal lance, for granules from 2 up to 4 mm Ø
    300 mm
    2 - 4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 180,00 €
    2 180,00 €
    1 * 1 ST
       2 180,00 €
  • 300-0194
    Sampler, stainless steel V4A, with vertical lance, for granules from 2 up to 4 mm Ø
    Sampler, stainless steel V4A, with vertical lance, for granules from 2 up to 4 mm Ø
    600 mm
    2 - 4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 340,00 €
    2 340,00 €
    1 * 1 ST
       2 340,00 €
  • 300-0196
    Sampler, stainless steel V4A, with vertical lance, for granules from 2 up to 4 mm Ø
    Sampler, stainless steel V4A, with vertical lance, for granules from 2 up to 4 mm Ø
    900 mm
    2 - 4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 450,00 €
    2 450,00 €
    1 * 1 ST
       2 450,00 €
  • 300-0197
    Sampler, stainless steel V4A, with vertical lance for granules from 2 up to 4 mm Ø
    Sampler, stainless steel V4A, with vertical lance for granules from 2 up to 4 mm Ø
    1200 mm
    2 - 4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 720,00 €
    2 720,00 €
    1 * 1 ST
       2 720,00 €
Additional spiral screws for powders
Product Information
  • Length
    Ø
    Availability
    Price
  • BURK5302-0316
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 300 mm
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 338,00 €
    338,00 €
    1 * 1 ST
       338,00 €
  • BURK5302-0616
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 600 mm
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 600 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 368,00 €
    368,00 €
    1 * 1 ST
       368,00 €
  • BURK5302-0916
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 900 mm
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 900 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 404,00 €
    404,00 €
    1 * 1 ST
       404,00 €
  • BURK5302-1216
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 1200 mm
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 1200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 435,00 €
    435,00 €
    1 * 1 ST
       435,00 €
Additional spiral screws for granules
Product Information
  • Length
    Ø
    Availability
    Price
  • BURK5302-0317
    Spiral screw for PowderProof® for granules 2 - 4 mm Ø, for 300 mm
    Spiral screw for PowderProof® for granules 2 - 4 mm Ø, for 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 338,00 €
    338,00 €
    1 * 1 ST
       338,00 €
  • BURK5302-0617
    Spiral screw for PowderProof® for granules 2 - 4 mm Ø, for 600 mm
    Spiral screw for PowderProof® for granules 2 - 4 mm Ø, for 600 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 368,00 €
    368,00 €
    1 * 1 ST
       368,00 €
  • BURK5302-0917
    Spiral screw for PowderProof® for granules 2 - 4 mm Ø, for 900 mm
    Spiral screw for PowderProof® for granules 2 - 4 mm Ø, for 900 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 404,00 €
    404,00 €
    1 * 1 ST
       404,00 €
  • BURK5302-1217
    Spiral screw for PowderProof® for granules 2 - 4 mm Ø, for 1200 mm
    Spiral screw for PowderProof® for granules 2 - 4 mm Ø, for 1200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 435,00 €
    435,00 €
    1 * 1 ST
       435,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Transport case
    Transport case
    312-1141
    Log in for availability
    1 * 1 ST - 135,00 €
Product Information
  • Catalog #
    Capacity
    Thread
    Material
    Availability
    Price
  • 300-0174
    500 ml
    50 mm
    LDPE
    Log in for availability
    1 * 1 ST - 3,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Packaging bags with closure (120×170 mm), 400 ml
    Packaging bags with closure (120×170 mm), 400 ml
    113-5414
    Log in for availability
    1 * 100 ST - 13,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Clamp for sampling bags, Ø 25 mm
    Clamp for sampling bags, Ø 25 mm
    BURK5338-0001
    Log in for availability
    1 * 1 ST - 20,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Basket for sample bottles 500 ml, Ø 84 mm, including holding ring
    Basket for sample bottles 500 ml, Ø 84 mm, including holding ring
    BURK5302-2055
    Log in for availability
    1 * 1 ST - 107,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Basket for sample bottles 1000 ml, Ø 100 mm, including holding ring
    Basket for sample bottles 1000 ml, Ø 100 mm, including holding ring
    BURK5302-2105
    Log in for availability
    1 * 1 ST - 135,00 €