Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Lieferant: Simport Scientific
Zertifikate
Über diesen Artikel
Acetal polymer. Sleeved cassettes especially made to be used with Thermo Fisher printers. Cassettes stacked with tape should be used with Leica and Sakura inkjet printers.
- The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage
- Keep specimens safely submerged
- Totally resistant to the chemical action of solvents used in histology laboratories
Frequently Bought Together