Order Entry
Austria
ContactUsLinkComponent
Petri dishes, square with grid
Petri dishes, square with grid
Catalog # SIMPD210-16
Supplier:  Simport Scientific
undefined
Petri dishes, square with grid
Catalog # SIMPD210-16
Supplier:  Simport Scientific

Specifications

  • Ø:
    90 mm
  • Packed:
    50 bags with 10 pieces
  • Sterility:
    Sterile
  • Version:
    With four vents, square with grid
  • Height:
    15 mm
  • Seq:
    01
  • Pk:
    500

Specifications

About this item

PS, sterile. Free from optical distortion, these dishes are ideal for phage typing, susceptibility testing, plate counts, and probe assays. Each 13 mm grid is marked numerically in one direction and alphabetically in the other. This dish has four venting ribs into the underside of the lid to prevent condensation build-up.