Blood collecting sets, Vacutainer® Safety-Lok™
Lieferant: BD Medical
Zertifikate
Über diesen Artikel
Blood collection sets consist of a needle and a multi-sample Luer adapter, connected by several length of tubing. A Safety-Lok protective shield reduces needlestick risk.
- Needles available with a pre-attached holder
- Luer adapter allows specimen collection from a 'closed system'
- Translucent shield shows blood flashback clearly and locks retracted needle into place after use
Certifications: Safety-Lok shields meet current OSHA safety standards for engineering controls.
Ordering information: Additional Luer adapters are available.
Delivery: Supplied sterile, slim and easy to handle, with larger wing for improved positioning.
Frequently Bought Together