Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample cups, Fibrin

Sample cups, Fibrin

For Fibrin analysers, 1,3 ml, Bag of 1000, case of 10000, 11,9 mm
Catalog # SIMPB720-40
Supplier:   Simport Scientific
Sample cups, Fibrin
Catalog # SIMPB720-40
Supplier:  Simport Scientific
Sample cups, Fibrin
Sample cups, Fibrin

For Fibrin analysers, 1,3 ml, Bag of 1000, case of 10000, 11,9 mm

Price
59,40 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

PE, translucent.

  • Good chemical resistance
  • Good temperature resistance

Precision molded for constant volume and uniform heat transfer.

Specifications
  • Ø opening:
    11,9 mm
  • Packed:
    Bag of 1000, case of 10000
  • Capacity:
    1,3 ml
  • Description:
    For Fibrin analysers
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.