Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Dissolved oxygen bottles
Supplier:   Brand
Dissolved oxygen bottles
Supplier:  Brand
Certificates
Dissolved oxygen bottles
2 Options Available Below
Product Details & Documents
Soda-lime glass.
  • Solid, obliquely cut standard-ground glass stopper can be secured with a fastening clip
  • Each bottle is calibrated with its own stopper; hence, stoppers and bottles are not interchangeable
  • Each bottle and its stopper is marked with a unique matching identification number
For the determination of oxygen dissolved water. The measured volume is indicated to ± 0,01 ml. White labelling area.
Product Family Options
Product Information
  • Capacity
    NS stopper
    Availability
    Price
  • 528-1303
    Oxygen bottles acc. to Winkler, with stopper
    Oxygen bottles acc. to Winkler, with stopper
    100 - 150 ml
    14/23
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,60 €
    28,60 €
    1 * 1 ST
       28,60 €
  • BRND386048
    Oxygen bottles acc. to Winkler, with stopper
    Oxygen bottles acc. to Winkler, with stopper
    250 - 300 ml
    19/26
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 64,80 €
    64,80 €
    1 * 2 ST
       64,80 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Fastening clip for dissolved oxygen bottles, winkler pattern with stopper NS 19/26
    Fastening clip for dissolved oxygen bottles, winkler pattern with stopper NS 19/26
    528-1311
    Log in for availability
    1 * 1 ST - 7,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Fastening clip for dissolved oxygen bottles, winkler pattern with stopper NS 14/23
    Fastening clip for dissolved oxygen bottles, winkler pattern with stopper NS 14/23
    BRND386138
    Log in for availability
    1 * 1 ST - 6,60 €