Laboratory bottles, narrow neck, with screw cap and thread
Bottles are made of DURAN® borosilicate glass, with DIN thread, blue PP screw caps and pouring rings.
- Studded base for stability
- Resistant to abrasions
- With graduation and inscription label
: ISO 4796
Frequently Bought Together