Order Entry
Austria
ContactUsLinkComponent
 
2,3,4,6-Tetrachlorophenol
Catalog # EHERC17374600
Supplier:  EHRENSTORFER
CAS Number:  
2,3,4,6-Tetrachlorophenol
Catalog # EHERC17374600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :231.89 g/mol
Boiling Pt:268 °C (1013 hPa)
Melting Pt:70 °C
Density:1.839 g/cm³ (20 °C)
EINECS:200-402-8
UN:2020
ADR:6.1,III
 :GHS06
 :GHS09
CAS Number:58-90-2
Formula:C₆H₂Cl₄O