Orders LinkContactUsLinkComponent
Buckets, Round, PP
Buckets, Round, PP
Supplier:   BURKLE
Buckets, Round, PP
Supplier:  BURKLE
Certificates
Buckets, Round, PP
Buckets, Round, PP
2 Options Available Below
Product Details & Documents
Made of PP, translucent.
  • With handle
  • With spout
  • With contents scale, graduation 1000 ml
  • Sterilisable
Bucket for transporting, decanting and mixing. Safe emptying with wide spout.
Product Family Options
Product Information
  • Colour
    Capacity
    Ø
    Height
    Availability
    Price
  • Transparent
    12 L
    310 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
  • Transparent
    15 L
    300 mm
    340 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,70 €
    46,70 €
    1 * 1 ST
       46,70 €