Adapters for distilling receivers, Bredt, bent
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Bent adapters (105°), with two NS sockets, vacuum connection: GL 18.
Frequently Bought Together