Drying tubes with standard ground cone, bent, DURAN®
DURAN® borosilicate glass 3.3. clear. Angled at 75° with ground joint cone.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.
Frequently Bought Together