Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue embedding cassettes, mega Type

Tissue embedding cassettes, mega Type

Supplier:   KARTELL
Tissue embedding cassettes, mega Type
Supplier:  KARTELL
Certificates
Tissue embedding cassettes, mega Type
Tissue embedding cassettes, mega Type

5 Options Available Below

Product Details & Documents

Material: POM. Tissue embedding cassettes Mega Type for samples with big dimensions. Dimension: 28×40×13,6 mm.

Product Family Options

Product Information

  • Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 396,00 €
    396,00 €
    1 * 1.000 ST
       396,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 396,00 €
    396,00 €
    1 * 1.000 ST
       396,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 396,00 €
    396,00 €
    1 * 1.000 ST
       396,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 396,00 €
    396,00 €
    1 * 1.000 ST
       396,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 396,00 €
    396,00 €
    1 * 1.000 ST
       396,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.