Tissue embedding cassettes, UNI type
Supplier: KARTELL
Jump to:
About this item
Tissue embedding cassettes for paraffin inclusions, Universal type, provided with clip on closure lid that can easily be removed. They are available in five colours. Dimensions: 28×40×6,8 mm.
6 Options Available Below
Product Details & Documents
Tissue embedding cassettes for paraffin inclusions, Universal type, provided with clip on closure lid that can easily be removed. They are available in five colours. Dimensions: 28×40×6,8 mm.
Product Family Options
Product Information
- ColourAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.631-9610 - Specifications:
More
Specifications
Cat. No.631-9611 - Specifications:
More
Specifications
Cat. No.631-9613 - Specifications:
More
Specifications
Cat. No.631-9614 - Specifications:
More
Specifications
Cat. No.631-9615 - Specifications:
More
Specifications
Cat. No.KART2921-10
Accessories
Product Information
- Catalog #AvailabilityPrice
- Tissue embedding sponge, blue