Vacuum filtering flasks, Büchner, with glass hose connection
Supplier: SciLabware
Certificates
About this item
PYREX®, borosilicate glass.
- For vacuum filtration
- Heavy walls provide increased mechanical strength
- 'Studded' base to minimise surface abrasion
Büchner filter flasks with integral glass side arm to accept 9 mm bore flexible tubing.
Certifications: Complies with ISO 6556
Frequently Bought Together