Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Clinical Diagnostic Supplies
Accessories for Blood Collection Tubes BD Microtainer®, BD Medical
Accessories for Blood Collection Tubes BD Microtainer®, BD Medical
BDAM368933
:
BD Medical
Accessories for Blood Collection Tubes BD Microtainer®, BD Medical
BDAM368933
:
BD Medical
Description:
Micro collection accessory, KFK312, extender for BD Microtainer® with BD Microgard™ closure, to increase the length to 75mm
Pk:
200