Order Entry
Austria
ContactUsLinkComponent
Pyromellitic acid for synthesis, Sigma-Aldrich®
Pyromellitic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Pyromellitic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    15 g/l (25 °C)
  • Melting Point:
    250 - 260 °C
  • Molar Mass:
    254.15 g/mol
  • Density:
    1.79 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together