
Pyromellitic acid 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89-05-4
:
Formula:C₁₀H₆O₈
MDL Number:MFCD00002471
:254.15 g/mol
Merck Index:14,8004
Boiling Pt:516 °C (1013 hPa)
Melting Pt:281…285 °C
Density:1.79 g/cm³ (20 °C)
EINECS:201-879-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together