ContactUsLinkComponent
Robotic tips for BioTek™ pipetting systems
Robotic tips for BioTek™ pipetting systems
  613-1887
   Low Manufacturing Impact
undefined
Robotic tips for BioTek™ pipetting systems
  613-1887
   Low Manufacturing Impact
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    150 µl
  • Seq:
    07
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Tip style:
    Filtered, low retention
  • For use with:
    BioTek™
  • Filter:
    +
  • Pk:
    960
These automation tips for BioTek™ pipetting systems undergo a rigorous 15-point quality control program to ensure consistency thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation.
  • Sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)