Orders LinkContactUsLinkComponent
BeefSteaker
BeefSteaker
BeefSteaker complete, 25 mm
Catalog # 312-0087
Supplier:   BURKLE
BeefSteaker
Catalog # 312-0087
Supplier:  BURKLE
BeefSteaker
BeefSteaker
BeefSteaker complete, 25 mm
Price
249,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
For taking bacteriological samples from the surfaces of dead animals in slaughterhouses and in butchers‘ shops. (cattle, pigs, sheep, goats and horses). The BeefSteaker is used to cut out a 25 mm round sample from the animal‘s skin. The stainless steel (1,4404) cutter allows the sample to be cut out easily.
  • Can be steam-sterilised or flamed
  • A hole in the crown prevents air compression and is used for ejecting the sample
The BeefSteaker can also be used for other sampling purposes (histology, medicine, food and semi-luxury articles, cheese factories, agriculture).
Documents
Specifications
  • Ø:
    25 mm
  • Description:
    BeefSteaker complete
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Transport case
    Transport case
    312-0088
    Log in for availability
    1 * 1 ST - 14,80 €