Order Entry
Austria
ContactUsLinkComponent
 
E.Z.N.A.® Formalin-Fixed, Paraffin-Embedded (FFPE) Tissue DNA Kits
 
E.Z.N.A.® Formalin-Fixed, Paraffin-Embedded (FFPE) Tissue DNA Kits

 

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    FFPE tissues
  • Target molecule:
    DNA

 

Frequently Bought Together