Orders LinkContactUsLinkComponent
Tubs, round
 :   BURKLE
Tubs, round
 :  BURKLE
Tubs, round
Tubs, round
Tubs, round, made of HDPE. For every purpose, with strong edge and practical carrying handles for safe transport. A lid can be delivered for every tub.
  • Temperature resistant from −30 to 70 °C
  • Suitable for food
  • High degree of chemical resistance
Product Family Options
  • Capacity
    Ext.Ø
    Height
  • 40 L
    370 mm
    440 mm
    1 * 1 ST
    1 * 1 ST - 71,80 €
    71,80 €
    1 * 1 ST
       71,80 €
  • 50 L
    370 mm
    555 mm
    1 * 1 ST
    1 * 1 ST - 85,50 €
    85,50 €
    1 * 1 ST
       85,50 €
  • 70 L
    460 mm
    535 mm
    1 * 1 ST
    1 * 1 ST - 103,00 €
    103,00 €
    1 * 1 ST
       103,00 €
  • 110 L
    460 mm
    710 mm
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
Accessories
  •  
    Description
  • BURK3482-0751
    Lids for drums, round, for 460 mm Ø, 75 litres capacity
    1 * 1 ST - 39,60 €
  •  
    Description
  • BURK3482-0401
    Lids for drums, round, for 370 mm Ø, 40 litres capacity
    1 * 1 ST - 17,50 €