Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes, Multipurpose

Storage Boxes, Multipurpose

Supplier:   BURKLE
Storage Boxes, Multipurpose
Supplier:  BURKLE
Certificates
Storage Boxes, Multipurpose
Storage Boxes, Multipurpose

3 Options Available Below

Product Details & Documents

Made of HDPE, these practical storage containers are ideal for safe transport of pourable or liquid contents.

  • With closed gripping edges
  • Can be stacked
  • Rounded inner edges and smooth inner surfaces for easy cleaning
  • Enables use in foodstuffs/hygiene sector
Product Family Options

Product Information

  • Capacity
    Colour
    W×D×H
    Material
    Availability
    Price
  • 29 l
    White
    660×450×130 mm
    HDPE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,30 €
    47,30 €
    1 * 1 ST
       47,30 €

  • 48 l
    White
    660×450×220 mm
    HDPE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,70 €
    66,70 €
    1 * 1 ST
       66,70 €

  • 94 l
    White
    660×450×410 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 101,00 €
    101,00 €
    1 * 1 ST
       101,00 €

Accessories

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Lid for universal storage containers, PS, white
    Lid for universal storage containers, PS, white
    BURK3418-0001
    - mm
    Log in for availability
    1 * 1 ST - 26,50 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.